KEY466133625 Key Organics / BIONET

1-methyl-4-[2-(4-nitrobenzoyl)-5-(trifluoromethyl)pyridin-3-yl]piperazine

  • MDL number MFCD00244644
  • Cas Number : 306977-48-0
  • Molecular Weight : 394.354

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8152

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12H-035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart