KEY466133638 Key Organics / BIONET

1-(4-chlorophenyl)-2-{[(4-methoxyphenyl)methyl]amino}-1-phenylethan-1-ol

  • MDL number MFCD01316762
  • Cas Number : 306977-76-4
  • Molecular Weight : 367.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8198

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12H-350S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart