KEY466133639 Key Organics / BIONET

ethyl 2-{2-[(E)-2-phenylethenyl]-1H-1,3-benzodiazol-1-yl}acetate

  • MDL number MFCD00215560
  • Cas Number : 1190919-79-9
  • Molecular Weight : 306.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Light sensitive
Shipping Temperature Light sensitive
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8220

Safety Information

signalword Warning
pcodes P410
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12H-530S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart