KEY466133641 Key Organics / BIONET

2-methoxy-6-(4-methoxyphenyl)pyridine-3-carbonitrile

  • MDL number MFCD00215275
  • Cas Number : 306977-92-4
  • Molecular Weight : 240.262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8227

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12H-537S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart