KEY466133655 Key Organics / BIONET

3-(4-fluorophenyl)-5-[(1Z)-1-[2-(2-fluorophenyl)hydrazin-1-ylidene]ethyl]-2,1-benzoxazole

  • MDL number MFCD00663806
  • Cas Number : 306978-26-7
  • Molecular Weight : 363.368

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8274

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12H-941_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart