KEY466133657 Key Organics / BIONET

ethyl 2-({4-oxo-4H-pyrido[1,2-a][1,3,5]triazin-2-yl}sulfanyl)propanoate

  • MDL number MFCD00243879
  • Cas Number : 306978-51-8
  • Molecular Weight : 279.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8307

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12J-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart