KEY466133663 Key Organics / BIONET

2-benzoyl-5,7-dichloro-3-methyl-1-benzofuran

  • MDL number MFCD00244009
  • Cas Number : 101081-68-9
  • Molecular Weight : 305.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8343

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12J-534S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart