KEY466133668 Key Organics / BIONET

2-(3-hydroxy-4-oxo-3,4-dihydroquinazolin-2-yl)benzoic acid

  • MDL number MFCD00975306
  • Cas Number : 37833-80-0
  • Molecular Weight : 282.255

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8353

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12J-560S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart