KEY466133672 Key Organics / BIONET

(E)-[4-(benzenesulfonyl)butan-2-ylidene][(4-nitrophenyl)methoxy]amine

  • MDL number MFCD00243409
  • Cas Number : 306979-45-3
  • Molecular Weight : 362.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8386

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12K-001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart