KEY466133690 Key Organics / BIONET

(1Z)-1-(dimethylamino)-4-(3-fluorophenoxy)pent-1-en-3-one

  • MDL number MFCD00243590
  • Cas Number : 1164520-33-5
  • Molecular Weight : 237.274

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8424

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12K-040_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart