KEY466133701 Key Organics / BIONET

methyl 6-[benzyl(methyl)amino]-5-cyano-2-methylpyridine-3-carboxylate

  • MDL number MFCD00793611
  • Cas Number : 306979-95-3
  • Molecular Weight : 295.342

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8455

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12K-322S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart