KEY466133704 Key Organics / BIONET

methyl 5-cyano-6-(2,6-dimethylmorpholin-4-yl)-2-methylpyridine-3-carboxylate

  • MDL number MFCD00793614
  • Cas Number : 306979-98-6
  • Molecular Weight : 289.335

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8458

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12K-325S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart