KEY466133713 Key Organics / BIONET

6-{[(4-chlorophenyl)sulfanyl]methyl}-2-phenyl-N-propylpyrimidin-4-amine

  • MDL number MFCD00793524
  • Cas Number : 306980-49-4
  • Molecular Weight : 369.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8524

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12K-525S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart