KEY466133729 Key Organics / BIONET

3-cyano-N-[3-cyano-4-(methylsulfanyl)phenyl]benzamide

  • MDL number MFCD00793924
  • Cas Number : 320421-53-2
  • Molecular Weight : 293.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12K-625S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart