KEY466133732 Key Organics / BIONET

N'-[(1E)-1-(2,4,5-trimethoxyphenyl)ethylidene]methoxycarbohydrazide

  • MDL number MFCD00793714
  • Cas Number : 320421-86-1
  • Molecular Weight : 282.296

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8641

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-007_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart