KEY466133733 Key Organics / BIONET

N-(butan-2-yl)-5-(1,2,3-thiadiazol-4-yl)thiophene-2-sulfonamide

  • MDL number MFCD00793716
  • Cas Number : 320421-88-3
  • Molecular Weight : 303.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8643

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart