KEY466133739 Key Organics / BIONET

2-[(2,4-dichlorophenyl)amino]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD00793732
  • Cas Number : 320422-00-2
  • Molecular Weight : 307.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8659

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart