KEY466133742 Key Organics / BIONET

(3E)-1-[(4-methylphenyl)methyl]-3-[2-(4-nitrophenyl)hydrazin-1-ylidene]-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD01314822
  • Cas Number : 320422-08-0
  • Molecular Weight : 386.411

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8668

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-306S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart