KEY466133744 Key Organics / BIONET

(3E)-3-[2-(3,4-dichlorophenyl)hydrazin-1-ylidene]-1-(propan-2-yl)-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD01314826
  • Cas Number : 320422-12-6
  • Molecular Weight : 348.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8672

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-310S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart