KEY466133751 Key Organics / BIONET

(3E)-3-{2-[2,6-dichloro-4-(trifluoromethyl)phenyl]hydrazin-1-ylidene}-1-methyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD01314834
  • Cas Number : 320422-23-9
  • Molecular Weight : 388.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8685

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-325S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart