KEY466133757 Key Organics / BIONET

2-{[(4-methylphenyl)methyl]sulfanyl}-4-phenyl-6-[(prop-2-en-1-yl)amino]pyrimidine-5-carbonitrile

  • MDL number MFCD01314841
  • Cas Number : 320422-40-0
  • Molecular Weight : 372.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8714

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-506S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart