KEY466133758 Key Organics / BIONET

2-{[(4-methylphenyl)methyl]sulfanyl}-4-phenyl-6-[(propan-2-yl)amino]pyrimidine-5-carbonitrile

  • MDL number MFCD01314843
  • Cas Number : 320422-42-2
  • Molecular Weight : 374.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8716

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-508S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart