KEY466133760 Key Organics / BIONET

3-[(3,4-dichlorophenyl)methoxy]-1-benzothiophene-2-carbonitrile

  • MDL number MFCD01314848
  • Cas Number : 320422-48-8
  • Molecular Weight : 334.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8723

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-516S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart