KEY466133761 Key Organics / BIONET

ethyl 1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carboxylate

  • MDL number MFCD01568267
  • Cas Number : 320422-49-9
  • Molecular Weight : 278.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8724

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-518S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart