KEY466133768 Key Organics / BIONET

3-[(4-bromophenyl)sulfanyl]-1-(4-chlorophenyl)propan-1-one

  • MDL number MFCD00098714
  • Cas Number : 270086-96-9
  • Molecular Weight : 355.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8752

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-555S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart