KEY466133769 Key Organics / BIONET

1-(4-chlorophenyl)-3-[(4-chlorophenyl)sulfanyl]propan-1-one

  • MDL number MFCD00100325
  • Cas Number : 270086-85-6
  • Molecular Weight : 311.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8753

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-556S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart