KEY466133775 Key Organics / BIONET

3-chloro-N'-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2,2-dimethylpropanehydrazide

  • MDL number MFCD00793400
  • Cas Number : 320422-72-8
  • Molecular Weight : 330.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8761

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart