KEY466133776 Key Organics / BIONET

(3Z)-3-[(dimethylamino)methylidene]-6-methyl-3,4-dihydro-2H-pyran-2,4-dione

  • MDL number MFCD00794119
  • Cas Number : 181277-21-4
  • Molecular Weight : 181.191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8763

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-704_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart