KEY466133782 Key Organics / BIONET

(3Z)-6-methyl-3-({[4-(propan-2-yl)phenyl]amino}methylidene)-3,4-dihydro-2H-pyran-2,4-dione

  • MDL number MFCD00794132
  • Cas Number : 477859-28-2
  • Molecular Weight : 271.316

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8785

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart