KEY466133784 Key Organics / BIONET

4-hydroxy-6-methyl-3-[(1E)-{[(4-methylphenyl)methoxy]imino}methyl]-2H-pyran-2-one

  • MDL number MFCD00794135
  • Cas Number : 477859-30-6
  • Molecular Weight : 273.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8788

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-736_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart