KEY466133786 Key Organics / BIONET

benzyl N-[1-(hydrazinecarbonyl)-2-(1H-imidazol-5-yl)ethyl]carbamate

  • MDL number MFCD00508155
  • Cas Number : 254980-19-3
  • Molecular Weight : 303.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8802

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-913_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart