KEY466133790 Key Organics / BIONET

1-(tert-butylamino)-3-[(2-methyl-1H-indol-4-yl)oxy]propan-2-ol

  • MDL number MFCD01315603
  • Cas Number : 23869-98-9
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8821

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-933_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart