KEY466133792 Key Organics / BIONET

ethyl 2,4-dioxo-3-phenyl-3-azabicyclo[3.1.0]hexane-6-carboxylate

  • MDL number MFCD01187976
  • Cas Number : 3120-09-0
  • Molecular Weight : 259.261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8824

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12L-937_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart