KEY466133800 Key Organics / BIONET

2-{3-[(3,4-dichlorophenyl)methyl]-2,4,5-trioxoimidazolidin-1-yl}acetic acid

  • MDL number MFCD00890606
  • Cas Number : 128043-98-1
  • Molecular Weight : 331.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8842

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-018_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart