KEY466133807 Key Organics / BIONET

ethyl 2-{3-[(2-chlorophenyl)methyl]-2,4,5-trioxoimidazolidin-1-yl}acetate

  • MDL number MFCD01314619
  • Cas Number : 128043-66-3
  • Molecular Weight : 324.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8856

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-034_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart