KEY466133810 Key Organics / BIONET

4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]phenol

  • MDL number MFCD27924203
  • Cas Number : 161949-56-0
  • Molecular Weight : 273.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111 - 113
Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/402873

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-039_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart