KEY466133811 Key Organics / BIONET

N'-(3-methanesulfonyl-4-methyl-6-phenylpyridin-2-yl)-N'-methylcyclopropanecarbohydrazide

  • MDL number MFCD01315696
  • Cas Number : 320423-04-9
  • Molecular Weight : 359.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8863

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-302S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart