KEY466133813 Key Organics / BIONET

(2Z)-3-[2-(ethylsulfanyl)-1-methyl-1H-imidazol-5-yl]-2-phenylprop-2-enenitrile

  • MDL number MFCD01315699
  • Cas Number : 320423-14-1
  • Molecular Weight : 269.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8878

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-320S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart