KEY466133825 Key Organics / BIONET

2-[(3,5-dichlorophenyl)carbamoyl]-2-methoxyacetic acid

  • MDL number MFCD04124497
  • Cas Number : 145592-20-7
  • Molecular Weight : 278.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8917

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-535S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart