KEY466133826 Key Organics / BIONET

1-{6-methyl-[1,2,4]triazolo[3,2-b][1,3]thiazol-5-yl}ethyl N-(4-methylbenzenesulfonyl)carbamate

  • MDL number MFCD02082589
  • Cas Number : 477859-66-8
  • Molecular Weight : 380.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-561S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart