KEY466133829 Key Organics / BIONET

methyl 5-chloro-4-[(1E)-{[(4-methylphenyl)methoxy]imino}methyl]-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD02102317
  • Cas Number : 477709-62-9
  • Molecular Weight : 383.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8937

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-573S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart