KEY466133830 Key Organics / BIONET

methyl 5-chloro-4-[(1E)-(hydroxyimino)methyl]-1-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD01444168
  • Cas Number : 320423-32-3
  • Molecular Weight : 279.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8939

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-575S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart