KEY466133833 Key Organics / BIONET

6-amino-4-(4-methoxyphenyl)-3-methyl-1-phenyl-1H,4H-pyrano[2,3-c]pyrazole-5-carbonitrile

  • MDL number MFCD00565127
  • Cas Number : 53316-60-2
  • Molecular Weight : 358.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8946

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart