KEY466133836 Key Organics / BIONET

2-{2-[(difluoromethyl)sulfanyl]pyrimidin-4-yl}phenol

  • MDL number MFCD01315010
  • Cas Number : 477859-75-9
  • Molecular Weight : 254.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8954

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-719_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart