KEY466133837 Key Organics / BIONET

2-(benzenesulfonyl)-3-phenyl-2,5-dihydro-1,2-oxazol-5-one

  • MDL number MFCD01315606
  • Cas Number : 320423-38-9
  • Molecular Weight : 301.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8961

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-901_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart