KEY466133845 Key Organics / BIONET

4-[(4-methylphenyl)methoxy]-N'-[(1E,2Z)-3-phenylprop-2-en-1-ylidene]benzohydrazide

  • MDL number MFCD01568774
  • Cas Number : 320423-65-2
  • Molecular Weight : 370.452

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9031

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-079_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart