KEY466133877 Key Organics / BIONET

methyl 4-({6-[4-(methoxycarbonyl)phenoxy]hexyl}oxy)benzoate

  • MDL number MFCD02081993
  • Cas Number : 121595-36-6
  • Molecular Weight : 386.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-208_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart