KEY466133889 Key Organics / BIONET

4-hydroxy-N-(2-methoxyphenyl)-1-methyl-2,2-dioxo-1H-2lambda6-thieno[3,2-c][1,2]thiazine-3-carbothioamide

  • MDL number MFCD01569122
  • Cas Number : 320423-79-8
  • Molecular Weight : 382.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9134

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-309S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart