KEY466133891 Key Organics / BIONET

3-{[(2-aminophenyl)sulfanyl]methyl}-2H,3H,5H-[1,3]thiazolo[2,3-b]quinazolin-5-one

  • MDL number MFCD02082109
  • Cas Number : 477860-22-3
  • Molecular Weight : 341.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9142

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-324S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart