KEY466133906 Key Organics / BIONET

2-amino-4-(4-nitrophenyl)-5-oxo-5,6,7,8-tetrahydro-4H-chromene-3-carbonitrile

  • MDL number MFCD00551384
  • Cas Number : 156176-90-8
  • Molecular Weight : 311.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9180

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/12N-372S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart